Learn More
Cayman Chemical THUNDRHumn IL2 TRFRET B 2400
SDP
Catalog No. 502893364
Encompass_Preferred
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-2 in cell culture supernatants
Catalog No. 50-289-3364 Supplier Cayman Chemical Supplier No. 5002312400 WELL
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-2 in cell culture supernatants