Learn More
Cayman Chemical THUNDRHumn IL2 TRFRET B 4800
SDP
Catalog No. 502892494
Encompass_Preferred
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-2 in cell culture supernatants
Catalog No. 50-289-2494 Supplier Cayman Chemical Supplier No. 5002314800 WELL
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-2 in cell culture supernatants