Learn More
Cayman Chemical THUNDRHumn IL2 TRFRET B 9600
SDP
Catalog No. 502893077
Encompass_Preferred
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-2 in cell culture supernatants
Catalog No. 50-289-3077 Supplier Cayman Chemical Supplier No. 5002319600 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-2 in cell culture supernatants