Learn More
Cayman Chemical THUNDRHumn IL6 TRFRET B 480Wel
SDP
Catalog No. 502893466
Encompass_Preferred
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-6 in cell culture supernatants
Catalog No. 50-289-3466 Supplier Cayman Chemical Supplier No. 500232480 WELL
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-6 in cell culture supernatants