Learn More
Cayman Chemical THUNDRHumn IL8 TRFRET B 4800
SDP
Catalog No. 502895364
Encompass_Preferred
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-8 (CXCL8; CGP-1; NAP-1) in cell culture supernatants
Catalog No. 50-289-5364 Supplier Cayman Chemical Supplier No. 5002334800 WELL
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-8 (CXCL8; CGP-1; NAP-1) in cell culture supernatants