Learn More
Cayman Chemical THUNDRHumn IL8 TRFRET B 480Wel
SDP
Catalog No. 502894445
Encompass_Preferred
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-8 (CXCL8; CGP-1; NAP-1) in cell culture supernatants
Catalog No. 50-289-4445 Supplier Cayman Chemical Supplier No. 500233480 WELL
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-8 (CXCL8; CGP-1; NAP-1) in cell culture supernatants