Learn More
Cayman Chemical THUNDRHumn InsulIn TRFRE 2400
SDP
Catalog No. 502893321
Encompass_Preferred
A sandwich TR-FRET immunoassay for the quantitative measurement of human Insulin in cell culture supernatants
Catalog No. 50-289-3321 Supplier Cayman Chemical Supplier No. 5003052400 WELL
A sandwich TR-FRET immunoassay for the quantitative measurement of human Insulin in cell culture supernatants