Learn More
Cayman Chemical THUNDRHumn InsulIn TRFRE 480
SDP
Catalog No. 502893127
Encompass_Preferred
A sandwich TR-FRET immunoassay for the quantitative measurement of human Insulin in cell culture supernatants
Catalog No. 50-289-3127 Supplier Cayman Chemical Supplier No. 500305480 WELL
A sandwich TR-FRET immunoassay for the quantitative measurement of human Insulin in cell culture supernatants