Learn More
Cayman Chemical THUNDRHumn InsulIn TRFRE 4800
SDP
Catalog No. 502895097
Encompass_Preferred
A sandwich TR-FRET immunoassay for the quantitative measurement of human Insulin in cell culture supernatants
Catalog No. 50-289-5097 Supplier Cayman Chemical Supplier No. 5003054800 WELL
A sandwich TR-FRET immunoassay for the quantitative measurement of human Insulin in cell culture supernatants