Learn More
Cayman Chemical THUNDRHumn InsulIn TRFRE 9600
SDP
Catalog No. 502892999
Encompass_Preferred
A sandwich TR-FRET immunoassay for the quantitative measurement of human Insulin in cell culture supernatants
Catalog No. 50-289-2999 Supplier Cayman Chemical Supplier No. 5003059600 WELL
Add to Cart
Add to Cart
Request a Quote
A sandwich TR-FRET immunoassay for the quantitative measurement of human Insulin in cell culture supernatants