Learn More
Cayman Chemical THUNDRHumn InsulIn TRFRE 96Wel
SDP
Catalog No. 502895766
Encompass_Preferred
A sandwich TR-FRET immunoassay for the quantitative measurement of human Insulin in cell culture supernatants
Catalog No. 50-289-5766 Supplier Cayman Chemical Supplier No. 50030596 WELL
A sandwich TR-FRET immunoassay for the quantitative measurement of human Insulin in cell culture supernatants