Learn More
Cayman Chemical THUNDRHumn TNFa TRFRET B 2400
SDP
Catalog No. 502894020
Encompass_Preferred
Sandwich TR-FRET immunoassay for the quantitative measurement of human TNFα in cell culture supernatants
Catalog No. 50-289-4020 Supplier Cayman Chemical Supplier No. 5002342400 WELL
Sandwich TR-FRET immunoassay for the quantitative measurement of human TNFα in cell culture supernatants