Learn More
Cayman Chemical THUNDRHumn TNFa TRFRET B 4800
SDP
Catalog No. 502893226
Encompass_Preferred
Sandwich TR-FRET immunoassay for the quantitative measurement of human TNFα in cell culture supernatants
Catalog No. 50-289-3226 Supplier Cayman Chemical Supplier No. 5002344800 WELL
Sandwich TR-FRET immunoassay for the quantitative measurement of human TNFα in cell culture supernatants