Learn More
Cayman Chemical THUNDRHumn TNFa TRFRET B 9600
SDP
Catalog No. 502893155
Encompass_Preferred
Sandwich TR-FRET immunoassay for the quantitative measurement of human TNFα in cell culture supernatants
Catalog No. 50-289-3155 Supplier Cayman Chemical Supplier No. 5002349600 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for the quantitative measurement of human TNFα in cell culture supernatants