Learn More
Cayman Chemical THUNDRHumn TNFa TRFRET B 96Wel
SDP
Catalog No. 502893234
Encompass_Preferred
Sandwich TR-FRET immunoassay for the quantitative measurement of human TNFα in cell culture supernatants
Catalog No. 50-289-3234 Supplier Cayman Chemical Supplier No. 50023496 WELL
Sandwich TR-FRET immunoassay for the quantitative measurement of human TNFα in cell culture supernatants