Learn More
Cayman Chemical THUNDRMouse IL2 TRFRET B 2400
SDP
Catalog No. 502895588
Encompass_Preferred
A sandwich TR-FRET immunoassay for the quantitative measurement of mouse IL-2 in cell culture supernatants
Catalog No. 50-289-5588 Supplier Cayman Chemical Supplier No. 5003112400 WELL
A sandwich TR-FRET immunoassay for the quantitative measurement of mouse IL-2 in cell culture supernatants