Learn More
Cayman Chemical THUNDRMouse IL2 TRFRET B 4800
SDP
Catalog No. 502895654
Encompass_Preferred
A sandwich TR-FRET immunoassay for the quantitative measurement of mouse IL-2 in cell culture supernatants
Catalog No. 50-289-5654 Supplier Cayman Chemical Supplier No. 5003114800 WELL
A sandwich TR-FRET immunoassay for the quantitative measurement of mouse IL-2 in cell culture supernatants