Learn More
Cayman Chemical THUNDRMouse IL2 TRFRET B 96Wel
SDP
Catalog No. 502896008
Encompass_Preferred
A sandwich TR-FRET immunoassay for the quantitative measurement of mouse IL-2 in cell culture supernatants
Catalog No. 50-289-6008 Supplier Cayman Chemical Supplier No. 50031196 WELL
A sandwich TR-FRET immunoassay for the quantitative measurement of mouse IL-2 in cell culture supernatants