Learn More
Cayman Chemical THUNDRPhosphoAKT pnT30 9600Wel
SDP
Catalog No. 502893236
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-AKT 1/2/3 (T308) in cell lysates
Catalog No. 50-289-3236 Supplier Cayman Chemical Supplier No. 5002109600 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-AKT 1/2/3 (T308) in cell lysates