Learn More
Cayman Chemical THUNDRPhosphoBTKY223 T 4800Wel
SDP
Catalog No. 502895964
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-BTK (Y223) in cell lysates
Catalog No. 50-289-5964 Supplier Cayman Chemical Supplier No. 5002404800 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-BTK (Y223) in cell lysates