Learn More
Cayman Chemical THUNDRPhosphoEGFRY1068 9600Wel
SDP
Catalog No. 502896000
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-EGFR (Y1068) in cell lysates
Catalog No. 50-289-6000 Supplier Cayman Chemical Supplier No. 5002189600 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-EGFR (Y1068) in cell lysates