Learn More
Cayman Chemical THUNDRPhosphoeIF4S209 4800Wel
SDP
Catalog No. 502893011
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-eIF4E (S209) in cell lysates
Catalog No. 50-289-3011 Supplier Cayman Chemical Supplier No. 5002464800 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-eIF4E (S209) in cell lysates