Learn More
Cayman Chemical THUNDRPhosphoeIF4S209 9600Wel
SDP
Catalog No. 502893638
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-eIF4E (S209) in cell lysates
Catalog No. 50-289-3638 Supplier Cayman Chemical Supplier No. 5002469600 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-eIF4E (S209) in cell lysates