Learn More
Cayman Chemical THUNDRPhosphoInsulIn Rece 2400
SDP
Catalog No. 502895677
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-Insulin Receptor β (Y1150/Y1151) in cell lysates
Catalog No. 50-289-5677 Supplier Cayman Chemical Supplier No. 5002092400 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-Insulin Receptor β (Y1150/Y1151) in cell lysates