Learn More
Cayman Chemical THUNDRPhosphoInsulIn Rece 4800
SDP
Catalog No. 502895844
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-Insulin Receptor β (Y1150/Y1151) in cell lysates
Catalog No. 50-289-5844 Supplier Cayman Chemical Supplier No. 5002094800 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-Insulin Receptor β (Y1150/Y1151) in cell lysates