Learn More
Cayman Chemical THUNDRPhosphop38a MAPK 2400Wel
SDP
Catalog No. 502895745
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-p38αβγ MAPK14/11/12 (T180/Y182) in cell lysates
Catalog No. 50-289-5745 Supplier Cayman Chemical Supplier No. 5002692400 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-p38αβγ MAPK14/11/12 (T180/Y182) in cell lysates