Learn More
Cayman Chemical THUNDRPhosphop38a MAPK 4800Wel
SDP
Catalog No. 502895746
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-p38αβγ MAPK14/11/12 (T180/Y182) in cell lysates
Catalog No. 50-289-5746 Supplier Cayman Chemical Supplier No. 5002694800 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-p38αβγ MAPK14/11/12 (T180/Y182) in cell lysates