Learn More
Cayman Chemical THUNDRPhosphop38a MAPK 9600Wel
SDP
Catalog No. 502893383
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-p38αβγ MAPK14/11/12 (T180/Y182) in cell lysates
Catalog No. 50-289-3383 Supplier Cayman Chemical Supplier No. 5002699600 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-p38αβγ MAPK14/11/12 (T180/Y182) in cell lysates