Learn More
Cayman Chemical THUNDRPhosphop38a MAPKT 4800
SDP
Catalog No. 502893361
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-p38α MAPK14 (T180/Y182) in cell lysates
Catalog No. 50-289-3361 Supplier Cayman Chemical Supplier No. 5002684800 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-p38α MAPK14 (T180/Y182) in cell lysates