Learn More
Cayman Chemical THUNDRPhosphop70 S6KT38 4800
SDP
Catalog No. 502893194
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-p70 S6K (T389) in cell lysates
Catalog No. 50-289-3194 Supplier Cayman Chemical Supplier No. 5002744800 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-p70 S6K (T389) in cell lysates