Learn More
Cayman Chemical THUNDRPhosphoRK12T202 4800Wel
SDP
Catalog No. 502895094
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-ERK1/2 (T202/Y204) in cell lysates.
Catalog No. 50-289-5094 Supplier Cayman Chemical Supplier No. 5002064800 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-ERK1/2 (T202/Y204) in cell lysates.