Learn More
Cayman Chemical THUNDRPhosphoRPS6S235S 4800Wel
SDP
Catalog No. 502895444
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-Ribosomal Protein S6 (S235/S236) in cell lysates.
Catalog No. 50-289-5444 Supplier Cayman Chemical Supplier No. 5002774800 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-Ribosomal Protein S6 (S235/S236) in cell lysates.