Learn More
Cayman Chemical THUNDRPhosphoSLP76S376 4800Wel
SDP
Catalog No. 502893075
Encompass_Preferred
The Phospho-SLP-76 (S376) assay kit is a homogeneous timeresolved Förster resonance energy transfer (TR-FRET) sandwich immunoassay
Catalog No. 50-289-3075 Supplier Cayman Chemical Supplier No. 5002214800 WELL
Add to Cart
Add to Cart
Request a Quote
The Phospho-SLP-76 (S376) assay kit is a homogeneous timeresolved Förster resonance energy transfer (TR-FRET) sandwich immunoassay