Learn More
Cayman Chemical THUNDRPhosphoSRCY419 T 4800Wel
SDP
Catalog No. 502895199
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-SRC (Y419) in cell lysates
Catalog No. 50-289-5199 Supplier Cayman Chemical Supplier No. 5002844800 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-SRC (Y419) in cell lysates