Learn More
Cayman Chemical THUNDRPhosphoSRCY419 T 9600Wel
SDP
Catalog No. 502892692
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-SRC (Y419) in cell lysates
Catalog No. 50-289-2692 Supplier Cayman Chemical Supplier No. 5002849600 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-SRC (Y419) in cell lysates