Learn More
Cayman Chemical THUNDRPhosphoSTAT1Y701 4800Wel
SDP
Catalog No. 502892693
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-STAT1 (Y701) in cell lysates.
Catalog No. 50-289-2693 Supplier Cayman Chemical Supplier No. 5002854800 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-STAT1 (Y701) in cell lysates.