Learn More
Cayman Chemical THUNDRPhosphoSTAT3Y705 9600Wel
SDP
Catalog No. 502895709
Encompass_Preferred
A sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-STAT3 (Y705) in cell lysates
Catalog No. 50-289-5709 Supplier Cayman Chemical Supplier No. 5002009600 WELL
Add to Cart
Add to Cart
Request a Quote
A sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-STAT3 (Y705) in cell lysates