Learn More
Cayman Chemical THUNDRPhosphoSTAT5Y694 9600Wel
SDP
Catalog No. 502895092
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-STAT5 (Y694) in cell lysates
Catalog No. 50-289-5092 Supplier Cayman Chemical Supplier No. 5002159600 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-STAT5 (Y694) in cell lysates