Learn More
Cayman Chemical THUNDRTotal BTK TRFRET Ce 2400
SDP
Catalog No. 502892382
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total BTK (both phosphorylated and non-phosphorylated) in cell lysates
Catalog No. 50-289-2382 Supplier Cayman Chemical Supplier No. 5002412400 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total BTK (both phosphorylated and non-phosphorylated) in cell lysates