Learn More
Cayman Chemical THUNDRTotal BTK TRFRET Ce 480
SDP
Catalog No. 502894022
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total BTK (both phosphorylated and non-phosphorylated) in cell lysates
Catalog No. 50-289-4022 Supplier Cayman Chemical Supplier No. 500241480 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total BTK (both phosphorylated and non-phosphorylated) in cell lysates