Learn More
Cayman Chemical THUNDRTotal BTK TRFRET Ce 4800
SDP
Catalog No. 502895951
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total BTK (both phosphorylated and non-phosphorylated) in cell lysates
Catalog No. 50-289-5951 Supplier Cayman Chemical Supplier No. 5002414800 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total BTK (both phosphorylated and non-phosphorylated) in cell lysates