Learn More
Cayman Chemical THUNDRTotal BTK TRFRET Ce 96
SDP
Catalog No. 502894329
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total BTK (both phosphorylated and non-phosphorylated) in cell lysates
Catalog No. 50-289-4329 Supplier Cayman Chemical Supplier No. 50024196 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total BTK (both phosphorylated and non-phosphorylated) in cell lysates