Learn More
Cayman Chemical THUNDRTotal BTK TRFRET Ce 9600
SDP
Catalog No. 502895965
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total BTK (both phosphorylated and non-phosphorylated) in cell lysates
Catalog No. 50-289-5965 Supplier Cayman Chemical Supplier No. 5002419600 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total BTK (both phosphorylated and non-phosphorylated) in cell lysates