Learn More
Cayman Chemical THUNDRTotal cMet TRFRET 4800
SDP
Catalog No. 502895597
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total c-Met in cell lysates
Catalog No. 50-289-5597 Supplier Cayman Chemical Supplier No. 5002634800 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total c-Met in cell lysates