Learn More
Cayman Chemical THUNDRTotal CREB TRFRET C 4800
SDP
Catalog No. 502895676
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total CREB in cell lysates
Catalog No. 50-289-5676 Supplier Cayman Chemical Supplier No. 5002444800 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total CREB in cell lysates