Learn More
Cayman Chemical THUNDRTotal eIF2a TRFRET 4800
SDP
Catalog No. 502895711
Encompass_Preferred
A sandwich TR-FRET immunoassay for semi-quantitative detection of Total EIF2α in cell lysates
Catalog No. 50-289-5711 Supplier Cayman Chemical Supplier No. 5002044800 WELL
Add to Cart
Add to Cart
Request a Quote
A sandwich TR-FRET immunoassay for semi-quantitative detection of Total EIF2α in cell lysates