Learn More
Cayman Chemical THUNDRTotal eIF2a TRFRET 96Wel
SDP
Catalog No. 502892376
Encompass_Preferred
A sandwich TR-FRET immunoassay for semi-quantitative detection of Total EIF2α in cell lysates
Catalog No. 50-289-2376 Supplier Cayman Chemical Supplier No. 50020496 WELL
A sandwich TR-FRET immunoassay for semi-quantitative detection of Total EIF2α in cell lysates