Learn More
Cayman Chemical THUNDRTotal eIF4E TRFRET 9600
SDP
Catalog No. 502893635
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total eIF4E (both phosphorylated and non-phosphorylated eIF4E) in cell lysates
Catalog No. 50-289-3635 Supplier Cayman Chemical Supplier No. 5002479600 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total eIF4E (both phosphorylated and non-phosphorylated eIF4E) in cell lysates