Learn More
Cayman Chemical THUNDRTotal GAPDH TRFRET 480
SDP
Catalog No. 502895488
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total GAPDH in cell lysates
Catalog No. 50-289-5488 Supplier Cayman Chemical Supplier No. 500249480 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total GAPDH in cell lysates