Learn More
Cayman Chemical THUNDRTotal GSK3 TRFRET 2400
SDP
Catalog No. 502893233
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total GSK3β in cell lysates
Catalog No. 50-289-3233 Supplier Cayman Chemical Supplier No. 5002522400 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total GSK3β in cell lysates